Phallacidin
Leverantör: Apollo Scientific
|
Fara
|
BIP0345-1MG
BIP0345-5MG
APOSBIP0345-1MGEA
3250
SEK
APOSBIP0345-1MG
APOSBIP0345-5MG
Phallacidin
Phallacidin
Formel:
C₃₇H₅₀N₈O₁₃S Storage Temperature: Omgivande |
CAS nummer:
26645-35-2 UN: 2811 ADR: 6.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...