Nalidixic acid
Leverantör: Apollo Scientific
Fara
BIN0134-100G BIN0134-25G
APOSBIN0134-100GEA 1300 SEK
APOSBIN0134-100G APOSBIN0134-25G
Nalidixic acid
Nalidixic acid
Active against gram-negative bacteria.
Formel: C₁₂H₁₂N₂O₃
MW: 232,24 g/mol
Smältpunkt: 225…230 °C
Storage Temperature: Omgivande
MDL Number: MFCD00006884
CAS nummer: 389-08-2
EINECS: 206-864-7