Guanidinhydroklorid 99,5%
Leverantör: Apollo Scientific
Varning
Synonyms: Guanadine hydrochloride, Aminoformamidine hydrochloride, Aminomethanamidine hydrochloride, Guanidine HCI, Guanidine chloride, Guanidinium chloride, Guanidine hydrochloride
BIMB2002-1KG BIMB2002-500G BIMB2002-5KG
APOSBIMB2002-1KGEA 395 SEK
APOSBIMB2002-1KG APOSBIMB2002-500G APOSBIMB2002-5KG
Guanidinhydroklorid 99,5%
Guanidinhydroklorid
Protein denaturant.
  • Toxic-LD50 of <500mg/Kg
Formel: NH₂C(=NH)NH₂·HCl
MW: 95,53 g/mol
Smältpunkt: 178…188 °C
Densitet: 1,344 g/cm³ (25 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00013026
CAS nummer: 50-01-1
EINECS: 200-002-3