Nitroblå tetrazolium
Leverantör: Apollo Scientific
Varning
Synonyms: 5,5'-Diphenyl-3,3'-bis(4-nitrophenyl)-2,2'-(3,3'-dimethoxybiphenyl-4,4'-ylene)ditetrazolium dichloride
BIMB1019-1G BIMB1019-250MG BIMB1019-5G
APOSBIMB1019-1GEA 620 SEK
APOSBIMB1019-1G APOSBIMB1019-250MG APOSBIMB1019-5G
Nitroblå tetrazolium
Nitroblå tetrazolium
NBT is used in conjunction with BCIP (BIMB1004) for colourimetric detection of alkaline phophatase activity in blotting, immuno-histochemical and cytochemistry techniques.
Formel: C₄₀H₃₀Cl₂N₁₀O₆
MW: 817,65 g/mol
Smältpunkt: 189 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00012159
CAS nummer: 298-83-9
EINECS: 206-067-4