5-Bromo-4-chloro-3-indolyl-β-D-glucoside 98%
Leverantör: Apollo Scientific
Varning
Synonyms: 5-Bromo-4-chloro-3-indolyl beta-D-Glucopyranoside, (2S,3R,4S,5S,6R)-2-((5-Bromo-4-chloro-1H-indol-3-yl)oxy)-6-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol, X-Glc, 5-Bromo-4-chloro-3-(beta-D-glucopyranosyloxy)indole
BIMB1002-1G BIMB1002-500MG BIMB1002-5G
APOSBIMB1002-1GEA 925 SEK
APOSBIMB1002-1G APOSBIMB1002-500MG APOSBIMB1002-5G
5-Bromo-4-chloro-3-indolyl-β-D-glucoside 98%
5-Bromo-4-chloro-3-indolyl-beta-D-glucoside
Histochemical substrate for B-Glucosidase.
Formel: C₁₄H₁₅BrClNO₆
MW: 408.629 g/mol
Smältpunkt: 249…251 °C
Storage Temperature: Frys
MDL Number: MFCD00063690
CAS nummer: 15548-60-4
EINECS: 239-603-0