D-Mannosamine hydrochloride
Leverantör: Apollo Scientific
Varning
Synonyms: Mannosamine HCl, (2S,3R,4S,5R)-2-Amino-3,4,5,6-tetrahydroxyhexanal hydrochloride, Mannosamine hydrochloride, 2-Amino-2-deoxy-D-mannose hydrochloride, D-Mannosamine HCl
BIM4501-1G BIM4501-500MG
APOSBIM4501-1GEA 430 SEK
APOSBIM4501-1G APOSBIM4501-500MG
D-Mannosamine hydrochloride
D-Mannosamine hydrochloride
Formel: C₆H₁₄ClNO₅
MW: 215,632 g/mol
Smältpunkt: 168 °C (dec.)
Storage Temperature: Kylskåp
MDL Number: MFCD00064557
CAS nummer: 5505-63-5
EINECS: 226-847-8