L-Metionin 99
Leverantör: Apollo Scientific
Synonyms: (S)-(-)-2-Amino-4-(methylthio)butyric acid, (S)-(-)-Methionine, L(-)-Methionine, (S)-(-)-2-Amino-4-(methylthio)butanoic acid, H-Met-OH, Methionine, (S)-2-Amino-4-(methylthio)butanoic acid
BIM0715-100G BIM0715-1KG BIM0715-500G
APOSBIM0715-100GEA 260 SEK
APOSBIM0715-100G APOSBIM0715-1KG APOSBIM0715-500G
L-Metionin 99
L-Metionin
Ph.Eur, Pyrogen free
Formel: C₅H₁₁NO₂S
MW: 149.21 g/mol
Kokpunkt: 278
Smältpunkt: 284 °C
Storage Temperature: Omgivande
MDL Number: MFCD00063097
CAS nummer: 63-68-3
EINECS: 200-562-9
Merck Index: 13,06004