Mitomycin

Leverantör: Apollo Scientific
Fara

Synonyms: Ametycin

BIM0133-10MG BIM0133-25MG
APOSBIM0133-10MGEA 1375 SEK
APOSBIM0133-10MG APOSBIM0133-25MG
Mitomycin
Mitomycin

Mitomycin C is a HIGHLY TOXIC antibiotic with antineoplastic properties.

Formel: C₁₅H₁₈N₄O₅
Smältpunkt: 360 °C
Storage Temperature: Kylskåp
CAS nummer: 50-07-7
EINECS: 200-008-6
UN: 3462
ADR: 6.1 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR