2-Morpholino-8-phenylchromone ≥99%
Leverantör: Apollo Scientific
Varning
Synonyms: LY294002, LY 294002, 2-Morpholino-8-phenyl-4H-chromen-4-one, 2-Morpholino-8-phenyl-4-oxo-4H-1-benzopyran
BIL2940-25MG BIL2940-5MG
APOSBIL2940-25MGEA 1550 SEK
APOSBIL2940-25MG APOSBIL2940-5MG
2-Morpholino-8-phenylchromone ≥99%
2-Morpholino-8-phenylchromone
Specific inhibitor (IC50 = 1.40 uM) of phosphatidylinositol 3-kinase (PI3K).
Formel: C₁₉H₁₇NO₃
MW: 307,35 g/mol
Smältpunkt: 180…187 °C
Storage Temperature: Frys
MDL Number: MFCD00270881
CAS nummer: 154447-36-6