L (+)-Lysin monohydroklorid ≥99%
Leverantör: Apollo Scientific
Varning
Synonyms: Lysine Hydrochloride, (S)-(+)-2,6-Diaminohexanoic acid monohydrochloride, L-Lysine monohydrochloride, H-Lys-OH·HCl, L-Lysine monohydrocholoride, L-Lysine Hydrochloride, (S)-(+)-Lysine monohydrochloride, (S)-(+)-2,6-Diaminocaproic acid monohydrochloride, H-Lys-OH.HCl
BIL0714-1KG BIL0714-500G
APOSBIL0714-1KGEA 855 SEK
APOSBIL0714-1KG APOSBIL0714-500G
L (+)-Lysin monohydroklorid ≥99%
L (+)-Lysin monohydroklorid
USP, Pyrogen free
Formel: H₂N(CH₂)₄CH(NH₂)CO₂H·HCl
MW: 182,65 g/mol
Smältpunkt: 263 °C
Densitet: 1,28 g/cm³ (25 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00064564
CAS nummer: 657-27-2