Aesculin sesquihydrate
Leverantör: Apollo Scientific
Varning
Synonyms: 6,7-Dihydroxycoumarin 6-glucoside sesquihydrate, Esculin sesquihydrate
BIK9005-100G BIK9005-250G BIK9005-50G
APOSBIK9005-100GEA 2500 SEK
APOSBIK9005-100G APOSBIK9005-250G APOSBIK9005-50G
Aesculin sesquihydrate
Aesculin sesquihydrate
Formel: C₁₅H₁₆O₉·1,5H₂O
MW: 367,31 g/mol
Smältpunkt: 195…198 °C
Storage Temperature: Omgivande
MDL Number: MFCD00149492
CAS nummer: 66778-17-4