KT5720
Leverantör: Apollo Scientific
|
Varning
|
BIK1003-1MG
BIK1003-100UG
APOSBIK1003-1MGEA
21850
SEK
APOSBIK1003-1MG
APOSBIK1003-100UG
KT5720
KT5720
Storage Temperature:
Frys |
CAS nummer:
108068-98-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...