Ionomycin free acid
Leverantör: Apollo Scientific
Varning
BII0421-10MG BII0421-5MG BII0421-25MG
APOSBII0421-10MGEA 4075 SEK
APOSBII0421-10MG APOSBII0421-5MG APOSBII0421-25MG
Ionomycin free acid
Ionomycin
Potent and selective calcium ionophore.
Formel: C₄₁H₇₂O₉
MW: 709.02 g/mol
Smältpunkt: 204…206 °C
Storage Temperature: Frys
MDL Number: MFCD06798385
CAS nummer: 56092-81-0