Histamine dihydrochloride 97%
Leverantör: Apollo Scientific
Varning
Synonyms: Histamine 2HCl, 4-(2-Aminoethyl)-1H-imidazole dihydrochloride, 2-(1H-Imidazol-4-yl)ethan-1-amine dihydrochloride
BIH0700-25G BIH0700-5G BIH0700-100G
APOSBIH0700-25GEA 685 SEK
APOSBIH0700-25G APOSBIH0700-5G APOSBIH0700-100G
Histamine dihydrochloride 97%
Histamine dihydrochloride
Formel: C₅H₁₁Cl₂N₃
MW: 184.07 g/mol
Smältpunkt: 249…252 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00012703
CAS nummer: 56-92-8
EINECS: 200-298-4