Eosin Y, pulver
Leverantör: Apollo Scientific
Varning
Synonyms: Acid Red 87, Tetrabromofluorescein, 2-(3,6-Dihydroxy-2,4,5,7-tetrabromoxanthen-9-yl)-benzoic acid, 2,4,5,7-Tetrabromofluorescein, Eosin yellowish, Solvent red 43, Bromofluorescein, Bromo acid J. TS, XL, or XX, CI 45380, Bronze Bromo ES, Spiro[isobenzofuran-1(3H),9'-[9H]xanthen]-3-one, 2',4',5',7'-tetrabromo-3',6'-dihydroxy-
BIGE1004-100G BIGE1004-25G
APOSBIGE1004-100GEA 445 SEK
APOSBIGE1004-100G APOSBIGE1004-25G
Eosin Y, pulver
Eosin Y
Formel: C₂₀H₈Br₄O₅
MW: 647.89 g/mol
Smältpunkt: 295
Storage Temperature: Omgivande
MDL Number: MFCD00036189
CAS nummer: 15086-94-9