3',4'-Dimethoxy-2'-hydroxyacetophenone
Leverantör: Apollo Scientific
Varning
Synonyms: Gallacetophenone 3',4'-dimethyl ether
BIG1002-5G BIG1002-25G BIG1002-1G
APOSBIG1002-5GEA 775 SEK
APOSBIG1002-5G APOSBIG1002-25G APOSBIG1002-1G
3',4'-Dimethoxy-2'-hydroxyacetophenone
3',4'-Dimethoxy-2'-hydroxyacetophenone
Formel: C₁₀H₁₂O₄
Storage Temperature: Omgivande
MDL Number: MFCD00016684
CAS nummer: 5396-18-9