Glycin 99
Leverantör: Apollo Scientific
Varning
Synonyms: Aminoacetic acid, Glycocoll, H-Gly-OH
BIG0709-1KG BIG0709-5KG
APOSBIG0709-1KGEA 770 SEK
APOSBIG0709-1KG APOSBIG0709-5KG
Glycin 99
Glycin
Component of tris-glycine-sds running buffers forpolyacrylamide gel electrophoresis.
  • Ph. Eur.
  • Pyrogen free
Formel: NH₂CH₂COOH
MW: 75.07 g/mol
Kokpunkt: 181
Smältpunkt: 240…245 °C (dec.)
Densitet: 1.595 g/cm³ (25 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00008131
CAS nummer: 56-40-6
EINECS: 200-272-2
Merck Index: 13,04504
Order Now
SPECIFIKATIONER
Environmentally Preferable