D (+)-Glukos, vattenfri
Leverantör: Apollo Scientific
Varning
Synonyms: Corn sugar, ᴅ-(+)-Glucose, Glucose, D-(+)-Dextrose, (2R,3S,4R,5R)-2,3,4,5,6-Pentahydroxyhexanal, Glucolin, Dextrose, D-Glucose
BIFS0119-1KG BIFS0119-2.5KG BIFS0119-500G
APOSBIFS0119-1KGEA 585 SEK
APOSBIFS0119-1KG APOSBIFS0119-2.5KG APOSBIFS0119-500G
D (+)-Glukos, vattenfri
D (+)-Glukos
Formel: C₆H₁₂O₆
MW: 180,16 g/mol
Smältpunkt: 153…156 °C
Storage Temperature: Omgivande
MDL Number: MFCD00148912
CAS nummer: 50-99-7
EINECS: 200-075-1