FITC (fluorescein-5-isothiocyanate, fluorescein isothiocyanate isomer I)
Leverantör: Apollo Scientific
Fara
Synonyms: 5-isothiocyanatofluorescein, Fluorescein 5(6)-isothiocyanate, Fluorescein Isothiocyanate, Isomer I
BIF4014-1G BIF4014-100MG BIF4014-2.5G
APOSBIF4014-1GEA 4275 SEK
APOSBIF4014-1G APOSBIF4014-100MG APOSBIF4014-2.5G
FITC (fluorescein-5-isothiocyanate, fluorescein isothiocyanate isomer I)
FITC (fluorescein-5-isothiocyanate, fluorescein isothiocyanate isomer I)
Formel: C₂₁H₁₁NO₅S
MW: 389,381 g/mol
Smältpunkt: >360 °C
Storage Temperature: Frys
MDL Number: MFCD00005063
CAS nummer: 3326-32-7
EINECS: 222-042-0