EDTA järn mononatriumsalt ≥99%
Leverantör: Apollo Scientific
Varning
Synonyms: Ethylenedinitrilotetra-acetic acid iron (III) sodium salt, Edetic acid iron (III) monosodium salt, Edetic acid iron (III) sodium salt, Ethylenediaminetetra-acetic acid iron (III) sodium salt, Iron (III) sodium ethylenediaminetetraacetate, Ethylenedinitrilotetra-acetic acid iron (III) monosodium salt, EDTA iron sodium, Sodium ferric ethylenediaminetetraacetate, Iron (III) monosodium ethylenediaminetetraacetate, Sodium feredetate, Ethylenediaminetetra-acetic acid iron (III) monosodium salt
BIE0509-100G BIE0509-1KG BIE0509-250G
APOSBIE0509-100GEA 225 SEK
APOSBIE0509-100G APOSBIE0509-1KG APOSBIE0509-250G
EDTA järn mononatriumsalt ≥99%
EDTA järn mononatriumsalt
Formel: C₁₀H₁₂FeN₂NaO₈
MW: 367,05 g/mol
Smältpunkt: 80 °C
Storage Temperature: Omgivande
MDL Number: MFCD00078215
CAS nummer: 15708-41-5
EINECS: 239-802-2