1,1'-Azobis(N,N-dimethylformamide)
Leverantör: Apollo Scientific
Varning
Synonyms: N1,N1,N2,N2-Tetramethyldiazene-1,2-dicarboxamide, Diamide
BID2040-5G BID2040-1G BID2040-500MG
APOSBID2040-5GEA 210 SEK
APOSBID2040-5G APOSBID2040-1G APOSBID2040-500MG
1,1'-Azobis(N,N-dimethylformamide)
1,1'-Azobis(N,N-dimethylformamide)
Possible thiol oxidising agent
  • Protein
Formel: C₆H₁₂N₄O₂
MW: 172,19 g/mol
Smältpunkt: 112…115 °C
Storage Temperature: Frys
MDL Number: MFCD00008318
CAS nummer: 10465-78-8
EINECS: 233-951-7