β-Cyclodextrin sulphobutyl ethers sodium salts (DS~10)
Leverantör: Apollo Scientific
Synonyms: SBE-β-CD, Betadex sulphobutyl ether sodium, Captisol
BICY1026-5G BICY1026-1G BICY1026-25G
APOSBICY1026-5GEA 5300 SEK
APOSBICY1026-5G APOSBICY1026-1G APOSBICY1026-25G
β-Cyclodextrin sulphobutyl ethers sodium salts (DS~10)
β-Cyclodextrin sulphobutyl ethers sodium salts
Formel: C₄₂H₇₀-nO₃₅.nNa.n(C₄H₈O₃S)
Storage Temperature: Omgivande
MDL Number: MFCD12032209
CAS nummer: 182410-00-0