β-Cyclodextrin sulphobutyl ethers sodium salts (DS~6)
Leverantör: Apollo Scientific
Synonyms: SBE-β-CD, Betadex sulphobutyl ether sodium, Captisol
BICY1025-25G BICY1025-5G BICY1025-100G
APOSBICY1025-25GEA 3425 SEK
APOSBICY1025-25G APOSBICY1025-5G APOSBICY1025-100G
β-Cyclodextrin sulphobutyl ethers sodium salts (DS~6)
β-Cyclodextrin sulphobutyl ethers sodium salts
Formel: C₄₂H₇₀-nO₃₅.nNa.n(C₄H₈O₃S)
Storage Temperature: Omgivande
MDL Number: MFCD12032209
CAS nummer: 182410-00-0