β-Cyclodextrin sulphobutyl ethers sodium salts (DS~4)
Leverantör: Apollo Scientific
Synonyms: SBE-β-CD, Betadex sulphobutyl ether sodium, Captisol
BICY1024-5G BICY1024-1G BICY1024-25G
APOSBICY1024-5GEA 5300 SEK
APOSBICY1024-5G APOSBICY1024-1G APOSBICY1024-25G
β-Cyclodextrin sulphobutyl ethers sodium salts (DS~4)
β-Cyclodextrin sulphobutyl ethers sodium salts
Formel: C₄₂H₇₀-nO₃₅.nNa.n(C₄H₈O₃S)
Storage Temperature: Omgivande
MDL Number: MFCD12032209
CAS nummer: 182410-00-0