4-Methoxyphenyl-2,3,4,6-tetra-O-acetyl-β-D-glucopyanoside 99% min
Leverantör: Apollo Scientific
Synonyms: (2R,3R,4S,5R,6S)-2-(Acetoxymethyl)-6-(4-methoxyphenoxy)tetrahydro-2H-pyran-3,4,5-triyl triacetate
BICL4135-25G BICL4135-5G
APOSBICL4135-25GEA 2400 SEK
APOSBICL4135-25G APOSBICL4135-5G
4-Methoxyphenyl-2,3,4,6-tetra-O-acetyl-β-D-glucopyanoside 99% min
4-Methoxyphenyl-2,3,4,6-tetra-O-acetyl-beta-D-glucopyanoside
Formel: C₂₁H₂₆O₁₁
Storage Temperature: Omgivande
MDL Number: MFCD06797142
CAS nummer: 14581-81-8