D-(+)-Glucose-[¹³C₆] 99% min (Isotope 13C 99% )
Leverantör: Apollo Scientific
Varning
Synonyms: (2R,3S,4R,5R)-2,3,4,5,6-Pentahydroxyhexanal-1,2,3,4,5,6-13C6, D-Glucose-13C6
BICL4109-1G BICL4109-500MG
APOSBICL4109-1GEA 24050 SEK
APOSBICL4109-1G APOSBICL4109-500MG
D-(+)-Glucose-[¹³C₆] 99% min (Isotope 13C 99% )
D-(+)-Glucose-[¹³C₆]
Formel: C₆H₁₂O₆
MW: 186,110 g/mol
Smältpunkt: 146…152 °C
Storage Temperature: Frys
MDL Number: MFCD00037544
CAS nummer: 110187-42-3