2,3:5,6-Di-O-isopropylidene-D-mannofuranose 99% min
Leverantör: Apollo Scientific
Synonyms: (+)-2,3:5,6-Di-O-isopropylidene-α-D-mannofuranose, 2,3:5,6-Di-O-isopropylidene-D-mannofuranose, (3aS,4S,6R,6aS)-6-((R)-2,2-Dimethyl-1,3-dioxolan-4-yl)-2,2-dimethyltetrahydrofuro[3,4-d][1,3]dioxol-4-ol, D-Mannose diacetonide
BICL4083-100G BICL4083-25G BICL4083-50G
APOSBICL4083-100GEA 16525 SEK
APOSBICL4083-100G APOSBICL4083-25G APOSBICL4083-50G
2,3:5,6-Di-O-isopropylidene-D-mannofuranose 99% min
2,3:5,6-Di-O-isopropylidene-D-mannofuranose
Formel: C₁₂H₂₀O₆
MW: 260,29 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00134206
CAS nummer: 14131-84-1