6,6'-Diazido-6,6'-dideoxytrehalose 98% min
Leverantör: Apollo Scientific
Varning
Synonyms: 6,6'-Diazido-6,6'-dideoxy-alpha,alpha-D-trehalose
BICL4068-250MG BICL4068-500MG BICL4068-100MG
APOSBICL4068-250MGEA 9225 SEK
APOSBICL4068-250MG APOSBICL4068-500MG APOSBICL4068-100MG
6,6'-Diazido-6,6'-dideoxytrehalose 98% min
6,6'-Diazido-6,6'-dideoxytrehalose
Formel: C₁₂H₂₀N₆O₉
MW: 392,33 g/mol
Storage Temperature: Omgivande
CAS nummer: 18933-88-5