N-Butyldeoxynojirimycin hydrochloride 99% min
Leverantör: Apollo Scientific
Synonyms: Miglustat HCl, (2R,3R,4R,5S)-1-Butyl-2-(hydroxymethyl)piperidine-3,4,5-triol hydrochloride
BICL4039-10MG BICL4039-25MG BICL4039-5MG
APOSBICL4039-10MGEA 4800 SEK
APOSBICL4039-10MG APOSBICL4039-25MG APOSBICL4039-5MG
N-Butyldeoxynojirimycin hydrochloride 99% min
N-Butyldeoxynojirimycin hydrochloride
Formel: C₁₀H₂₁NO₄·HCl
MW: 255,7 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00269940
CAS nummer: 210110-90-0