Cytidine-5'-triphosphoric acid disodium salt (CTP-Na2) dihydrate

Leverantör: Apollo Scientific
Varning

Synonyms: Sodium ((2R,3S,4R,5R)-5-(4-amino-2-oxopyrimidin-1(2H)-yl)-3,4-dihydroxytetrahydrofuran-2-yl)methyl dihydrogentriphosphate dihydrate, Cytidine-5'-Triphosphate disodium salt dihydrate

BIC5003-250MG BIC5003-500MG
APOSBIC5003-250MGEA 685 SEK
APOSBIC5003-250MG APOSBIC5003-500MG
Cytidine-5'-triphosphoric acid disodium salt (CTP-Na2) dihydrate
Cytidine-5'-triphosphoric acid disodium salt (CTP-Na2) dihydrate
Formel: C₉H₁₄N₃Na₂O₁₄P₃·2H₂O
MW: 527,13 g/mol (anhy)
Smältpunkt: 215…218 °C
Storage Temperature: Frys
MDL Number: MFCD00150807
CAS nummer: 81012-87-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR