Cysteaminium chloride 99%
Leverantör: Apollo Scientific
Varning
Synonyms: 2-MEA
BIC2510-100G BIC2510-25G
APOSBIC2510-100GEA 720 SEK
APOSBIC2510-100G APOSBIC2510-25G
Cysteaminium chloride 99%
Cysteaminium chloride
Formel: C₂H₇NS·HCl
MW: 113,61 g/mol
Smältpunkt: 66…68 °C
Densitet: 0,75 g/cm³ (25 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00012904
CAS nummer: 156-57-0
EINECS: 205-858-1
UN: 3335
ADR: 9,III