(3-((3-Cholamidopropyl) dimetylammonio)-2-hydroxi-1-propanesulfonate) ≥99.0%, Ultrapure
Leverantör: Apollo Scientific
Varning
Synonyms: (3-((3-Cholamidopropyl)dimethylammonio)-2-hydroxypropanesulphonate), CHAPSO
BIC0121-1G BIC0121-5G
APOSBIC0121-1GEA 515 SEK
APOSBIC0121-1G APOSBIC0121-5G
(3-((3-Cholamidopropyl) dimetylammonio)-2-hydroxi-1-propanesulfonate) ≥99.0%, Ultrapure
(3-((3-Cholamidopropyl) dimetylammonio)-2-hydroxi-1-propanesulfonate)
A non denaturing zwitterionic detergent similar to CHAPS(BIMB1085) with increased solubility.
Formel: C₃₂H₅₈N₂O₈S
MW: 630,89 g/mol
Smältpunkt: 184…186 °C
Storage Temperature: Omgivande
CAS nummer: 82473-24-3