Helvolic acid
Leverantör: Apollo Scientific
BIBR1050-1MG BIBR1050-5MG
APOSBIBR1050-1MGEA 1150 SEK
APOSBIBR1050-1MG APOSBIBR1050-5MG
Helvolic acid
Helvolic acid
Storage Temperature: Omgivande
CAS nummer: 29400-42-8