8-Aminopyrene-1,3,6-trisulphonic acid trisodium salt
Leverantör: Apollo Scientific
Varning
Synonyms: APTS
BIB6285-500MG BIB6285-100MG BIB6285-250MG
APOSBIB6285-500MGEA 13525 SEK
APOSBIB6285-500MG APOSBIB6285-100MG APOSBIB6285-250MG
8-Aminopyrene-1,3,6-trisulphonic acid trisodium salt
8-Aminopyrene-1,3,6-trisulphonic acid trisodium salt
Formel: C₁₆H₈NNa₃O₉S₃
MW: 523,40 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00674494
CAS nummer: 196504-57-1