Esculetin

Leverantör: Apollo Scientific
Varning

Synonyms: 6,7-Dihydroxycoumarin, 6,7-Dihydroxy-2H-chromen-2-one

BIB6051-10G BIB6051-25G
APOSBIB6051-10GEA 775 SEK
APOSBIB6051-10G APOSBIB6051-25G
Esculetin
Esculetin
Formel: C₉H₆O₄
MW: 178,14 g/mol
Smältpunkt: 270…273 °C (dec.)
Storage Temperature: Kylskåp
MDL Number: MFCD00006874
CAS nummer: 305-01-1
EINECS: 206-161-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR