DNA sodium salt (Deoxyribonucleic acid sodium salt) (from salmon testes)
Leverantör: Apollo Scientific
Synonyms: Thymonucleic acid sodium salt, DNA sodium salt
BIB6040-100G BIB6040-50G
APOSBIB6040-100GEA 10250 SEK
APOSBIB6040-100G APOSBIB6040-50G
DNA sodium salt (Deoxyribonucleic acid sodium salt) (from salmon testes)
DNA sodium salt (Deoxyribonucleic acid sodium salt)
Storage Temperature: Kylskåp
MDL Number: MFCD00130921
CAS nummer: 9007-49-2