6-Bromo-DL-tryptophan
Leverantör: Apollo Scientific
Synonyms: 2-Amino-3-(6-bromo-1H-indol-3-yl)propanoic acid, H-Trp(6-Br)-OH
BIB6019-1G BIB6019-250MG
APOSBIB6019-1GEA 4125 SEK
APOSBIB6019-1G APOSBIB6019-250MG
6-Bromo-DL-tryptophan
6-Bromo-DL-tryptophan
Formel: C₁₁H₁₁BrN₂O₂
MW: 283.12 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00055988
CAS nummer: 33599-61-0