Ethyl-5-(benzyloxy)-1H-indole-2-carboxylate
Leverantör: Apollo Scientific
Synonyms:
5-Benzyloxyindole-2-carboxylic acid ethyl ester, Ethyl 5-(benzyloxy)-1H-indole-2-carboxylate
BIB6003-25G
BIB6003-50G
APOSBIB6003-25GEA
15175
SEK
APOSBIB6003-25G
APOSBIB6003-50G
Ethyl-5-(benzyloxy)-1H-indole-2-carboxylate
Ethyl-5-(benzyloxy)-1H-indole-2-carboxylate
Formel:
C₁₈H₁₇NO₃ MW: 295,34 g/mol Smältpunkt: 161…165 °C Storage Temperature: Frys |
MDL Number:
MFCD00022702 CAS nummer: 37033-95-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...