Borrelidin

Leverantör: Apollo Scientific
Varning

BIB3043-10MG BIB3043-1MG BIB3043-5MG
APOSBIB3043-10MGEA 22675 SEK
APOSBIB3043-10MG APOSBIB3043-1MG APOSBIB3043-5MG
Borrelidin
Borrelidin

From Streptomyces sp.

Formel: C₂₈H₄₃NO₆
MW: 489.7 g/mol
Smältpunkt: 109…114 °C
Storage Temperature: Frys
CAS nummer: 7184-60-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR