Adenosine 5'-monophosphate monohydrate ≥96%
Leverantör: Apollo Scientific
Varning
Synonyms: A-5′-P, AMP, Adenosine-5'-monophosphoric acid monohydrate, 5′-AMP, 5′-Adenylic acid
BIB3002-25G BIB3002-100G
APOSBIB3002-25GEA 175 SEK
APOSBIB3002-25G APOSBIB3002-100G
Adenosine 5'-monophosphate monohydrate ≥96%
Adenosine 5'-monophosphate monohydrate
Formel: C₁₀H₁₄N₅O₇P·1H₂O
MW: 365,24 g/mol
Smältpunkt: 200 °C (dec.)
Storage Temperature: Kylskåp
MDL Number: MFCD00149360
CAS nummer: 18422-05-4