MES sodium salt
Leverantör: Apollo Scientific
Varning
Synonyms: MES-Na
BIB2014-100G BIB2014-500G
APOSBIB2014-100GEA 360 SEK
APOSBIB2014-100G APOSBIB2014-500G
MES sodium salt
MES sodium salt
Formel: C₆H₁₂NNaO₄S
MW: 217,22 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00065473
CAS nummer: 71119-23-8
EINECS: 275-203-2