β-(N-Morpholino)ethanesulphonic acid (MES) monohydrate
Leverantör: Apollo Scientific
Varning
Synonyms: MES monohydrate, 2-(N-Morpholino)ethanesulphonic acid monohydrate, 2-Morpholinoethanesulphonic acid hydrate
BIB2012-1KG BIB2012-250G BIB2012-500G
APOSBIB2012-1KGEA 3250 SEK
APOSBIB2012-1KG APOSBIB2012-250G APOSBIB2012-500G
β-(N-Morpholino)ethanesulphonic acid (MES) monohydrate
β-(N-Morpholino)ethanesulphonic acid (MES) monohydrate
Formel: C₆H₁₃NO₄S·1H₂O
MW: 213,25 g/mol
Smältpunkt: 308 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00149409
CAS nummer: 145224-94-8
EINECS: 224-632-3