n-Dodecyl-β-D-maltoside ≥99%, Ultrapure
Leverantör: Apollo Scientific
Varning
Synonyms: DDM
BIB2010-1G BIB2010-5G BIB2010-10G
APOSBIB2010-1GEA 310 SEK
APOSBIB2010-1G APOSBIB2010-5G APOSBIB2010-10G
n-Dodecyl-β-D-maltoside ≥99%, Ultrapure
n-Dodecyl-β-D-maltoside
Non ionic detergent used to extract proteins
  • Absorbance (1%) 260nm, 100mg/10ml, not more than 0.1
Formel: C₂₄H₄₆O₁₁
MW: 510.6 g/mol
Smältpunkt: 224…226 °C
Storage Temperature: Omgivande
MDL Number: MFCD00043012
CAS nummer: 69227-93-6