MOPS 99,5, ultrapure
Leverantör: Apollo Scientific
Varning
Synonyms: 3-Morpholinopropane-1-sulphonic acid, 3-(N-Morpholino)propanesulphonic acid
BIB2003-1KG BIB2003-250G BIB2003-500G
APOSBIB2003-1KGEA 2575 SEK
APOSBIB2003-1KG APOSBIB2003-250G APOSBIB2003-500G
MOPS 99,5, ultrapure
MOPS
Biological buffer - useful range 6.5-7.9
  • Heavy metals:<5 ppm
  • pKa @ 20°C: 7.20 + 0.2
Formel: C₇H₁₅NO₄S
MW: 209.27 g/mol
Smältpunkt: >277 °C
Flampunkt: 116 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00006183
CAS nummer: 1132-61-2
EINECS: 214-478-5
Merck Index: 13,06288