EDTA dinatriumsalt dihydrat 0,1 M volumetrisk lösning
Leverantör: Apollo Scientific
Varning
Synonyms: Ethylenediaminetetraacetic acid, disodium salt, dihydrate, Ethylenediaminetetraacetic acid disodium salt dihydrate, Kestranal® 2S dihydrate, Disodium dihydrogen ethylenediaminetetraacetate dihydrate, Ethylenedinitrilotetra-acetic acid disodium salt dihydrate, Ethylenediaminetetraacetic acid disodium dihydrate, Titriplex III, Ethylenediaminetetra-acetic acid disodium salt dihydrate, Kestranal® 2S 2 H2O, Sequestrene Na₂, Edetate disodium dihydrate, Edetic acid disodium salt dihydrate
BIA181670-1L BIA181670-5L
APOSBIA181670-1LEA 520 SEK
APOSBIA181670-1L APOSBIA181670-5L
EDTA dinatriumsalt dihydrat 0,1 M volumetrisk lösning
EDTA dinatriumsalt dihydrat
Formel: C₁₀H₁₄N₂Na₂O₈·2H₂O
MW: 372.24 g/mol
Smältpunkt: 255
Storage Temperature: Omgivande
MDL Number: MFCD00150037
CAS nummer: 6381-92-6
Merck Index: 13,03543