ABTS (Diammonium 2,2'-azinobis[3-ethyl-2,3-dihydrobenzothiazole-6-sulphonate])
Leverantör: Apollo Scientific
Varning
Synonyms: ABTS, Ammonium 2,2'-(hydrazine-1,2-diylidene)bis(3-ethyl-2,3-dihydrobenzo[d]thiazole-6-sulphonate), 2,2′-Azino-bis(3-ethylbenzthiazoline-6-sulphonic acid) diammonium salt
BIA1493-10G BIA1493-1G BIA1493-5G
APOSBIA1493-10GEA 4050 SEK
APOSBIA1493-10G APOSBIA1493-1G APOSBIA1493-5G
ABTS (Diammonium 2,2'-azinobis[3-ethyl-2,3-dihydrobenzothiazole-6-sulphonate])
ABTS (Diammonium 2,2'-azinobis[3-ethyl-2,3-dihydrobenzothiazole-6-sulphonate])
ABTS is a peroxidase substrate used in ELISA procedures.
Formel: C₁₈H₂₄N₆O₆S₄
MW: 548,68 g/mol
Smältpunkt: >300 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00010404
CAS nummer: 30931-67-0
EINECS: 250-396-6