3-Acetoxyindole

Leverantör: Apollo Scientific
Varning

Synonyms: 3-Indolyl Acetate, Indoxyl acetate, 3-Indoxyl acetate, 3-Indolyl acetate

BIA1216-10G BIA1216-1G BIA1216-5G
APOSBIA1216-10GEA 3250 SEK
APOSBIA1216-10G APOSBIA1216-1G APOSBIA1216-5G
3-Acetoxyindole
3-Acetoxyindole

Fluorogenic substrate for detaction of esterases.

Formel: C₁₀H₉NO₂
MW: 175,19 g/mol
Smältpunkt: 128…130 °C
Storage Temperature: Frys
MDL Number: MFCD00014561
CAS nummer: 608-08-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR