2-[4 (2-Hydroxietyl)-1-piperazinyl]-ethan sulfonsyra free acid ≥99.5%, Ultrapure
Leverantör: Apollo Scientific
Varning
Synonyms: HEPES free acid, N-(2-Hydroxyethyl)piperazine-N′-(2-ethanesulphonic acid), 4-(2-Hydroxyethyl)piperazin-1-ylethanesulphonic acid, N-2-Hydroxyethylpiperazine-N′-2-ethanesulphonic acid, 2-(4-(2-Hydroxyethyl)piperazin-1-yl)ethanesulphonic acid
BI8181-1KG BI8181-100G BI8181-250G
APOSBI8181-1KGEA 1100 SEK
APOSBI8181-1KG APOSBI8181-100G APOSBI8181-250G
2-[4 (2-Hydroxietyl)-1-piperazinyl]-ethan sulfonsyra free acid ≥99.5%, Ultrapure
HEPES
Biological buffer - useful range 6.8 - 8.2
  • Heavy metals: <1 ppm
  • pKa @ 20°C: 7.55 +/- 0.2
Formel: C₈H₁₈N₂O₄S
MW: 238.31 g/mol
Smältpunkt: 238…240 °C
Storage Temperature: Omgivande
MDL Number: MFCD00006158
CAS nummer: 7365-45-9
EINECS: 230-907-9
Merck Index: 13,04672
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4cdc09c866676149941b93b2cac8d453175059a2&vid=0b09e5b84ea34a806443c3071fad63e15e4e3433&prid=web&clid=SAPDAM