BIS-TRIS ≥99%
Leverantör: Apollo Scientific
Varning
Synonyms: 2,2-Bis(hydroxymethyl)-2,2',2''-nitrilotriethanol, Bis(2-hydroxyethyl)amino-tris(hydroxymethyl)methane (BIS-TRIS), 2-(Bis(2-hydroxyethyl)amino)-2-(hydroxymethyl)propane-1,3-diol, 2-[Bis(2-hydroxyethyl)amino]-2-(hydroxymethyl)propane-1,3-diol
BI3932-100G BI3932-25G BI3932-500G
APOSBI3932-100GEA 395 SEK
APOSBI3932-100G APOSBI3932-25G APOSBI3932-500G
BIS-TRIS ≥99%
BIS-TRIS
  • Assay (by titration) 98+%
  • Water (by Karl Fischer) < 1.0%
  • Heavy metals analysis < 0.0003%
  • Biological buffer - useful range 5.8-7.2
Formel: C₈H₁₉NO₅
MW: 209,24 g/mol
Smältpunkt: 102…105 °C
Storage Temperature: Omgivande
MDL Number: MFCD00002853
CAS nummer: 6976-37-0
EINECS: 230-237-7