3-(2,4-Difluorophenyl)pyrazole
Leverantör: Apollo Scientific
Varning
PC9993-250MG PC9993-1G
APOSPC9993-250MGEA 515 SEK
APOSPC9993-250MG APOSPC9993-1G
3-(2,4-Difluorophenyl)pyrazole
3-(2,4-Difluorophenyl)pyrazole
Formel: C₉H₆F₂N₂
MW: 180,154 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD01940429
CAS nummer: 474707-70-5